Pierce™ Dextran Desalting Columns, 5K MWCO, 5 ml, Thermo Scientific
Supplier: Invitrogen
43230 43233
PI43230PK 183.38 USD
PI43230 PI43233
Pierce™ Dextran Desalting Columns, 5K MWCO, 5 ml, Thermo Scientific
Chromatography Columns
Thermo Scientific Pierce Dextran Desalting Columns are ready-to-use, disposable, gel-filtration columns for separating proteins and other macromolecules from low-molecular weight buffer salts and reagents.
  • Stable in water, salt solutions, organic solvents and alkaline or acidic conditions
  • Excellent flow properties
  • Heat-stable
Pierce Dextran Desalting Columns (5K MWCO) are plastic columns that are pre-packed with crosslinked beaded dextran for protein desalting and buffer-exchange applications based on a molecular-weight cutoff (MWCO) of 5000.