Formic acid 99,5 LC-MS, Pierce™

Supplier: LIFE TECHNOLOGIES MS

Synonyms: Aminic Acid, Methanoic acid

28905 T85178-AD
PI28905PK 153.46 USD
PI28905 PI85178 10066-304
Formic acid 99,5 LC-MS, Pierce™
Formic acid

Thermo Scientific Pierce Formic Acid is high-purity solvent supplied in bottles as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.


Formic acid is a common component of reverse-phase mobile phases that provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing agent and it does not suppress MS ionization of polypeptides when used as a mobile-phase component.

Formula: HCO₂H
MW: 46.03
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8.2…8.4 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48 °C
MDL Number: MFCD00003297
CAS Number: 64-18-6
UN: 1779
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR