Octyl-β-D-thioglucopyranoside non-ionic detergent, Pierce™

Supplier: LIFE TECHNOLOGIES MS

Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

28351
PI28351EA 409.85 USD
PI28351
Octyl-β-D-thioglucopyranoside non-ionic detergent, Pierce™
Octyl β-D-thioglucopyranoside

Thermo Scientific Octylthioglucoside is a low-MW nonionic detergent that is effective for cell lysis and nondenaturing protein solubilization.


  • A low-molecular weight, nonionic detergent
  • Effective for a variety of cell lysis and protein solubilization methods
  • Unaffected by beta-glucosidase enzyme
  • More stable than octyl beta-glucoside (OG)
  • Can be removed from solution by dialysis
  • Optically clear; low absorbance at 280 nm


It is resistant to beta-D-glucoside glucohydrolase degradation. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8˚C.


Precaución: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR