Pierce™ Anti-GST Coated Plates, 8-Well Strip, Thermo Scientific
Supplier: Invitrogen
15144
PI15144EA 311.38 USD
PI15144
Pierce™ Anti-GST Coated Plates, 8-Well Strip, Thermo Scientific
Microplates Immunology Microplates and Strips
Thermo Scientific Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use
  • Detects GST fusion proteins down to 7.5ng/well
  • Binds native or denatured forms of GST; an alternative to glutathione-coated plates
  • Pre-blocked with SuperBlock blocking Buffer
  • Improved signal-to-noise ratio
Store plates at 4 °C and the unused plates can be stored in resealable bag.
Información para pedidos: Store plates at 4 °C. Store opened, unused plates in a resealable bag with desiccant at 4More Product Information