Biopsy Tray with Needles, CareFusion, BD Medical
Supplier: BD
BAK3411 BAK4511
PHBAK3411CS 1417.31 USD
PHBAK3411 PHBAK4511
Biopsy Tray with Needles, CareFusion, BD Medical
Trays/Dishes/Pots Instrument Trays/Dishes
Safe-T Plus and standard trays provide convenience and excellent procedural performance
The single-use, sterile trays include prepping, draping, local anesthetic, and procedural components needed to perform bone marrow biopsy procedures.