dATP, 25 µmol, Promega

Supplier: Promega Corporation

U1205
PAU1205EA 92.89 USD
PAU1205
dATP, 25 µmol, Promega
Nucleic Acid Reagents Nucleotides

High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.


  • High-Quality Nucleotides for Your Workflow
  • Greater than 99% triphosphate content
  • 100mM in water at pH 7.5


High-quality deoxynucleotide triphosphates (dNTPs) are critical for the success of many key procedures such as cDNA synthesis, sequencing and labeling. Promega dNTPs have greater than 99% triphosphate content and are provided at a concentration of 100mM in water at pH 7.5.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Concentration 100 mM
Storage Conditions -30°C to -10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR