4-CORE® Buffer Pack, Promega®
Supplier: Promega Corporation
R9921
PAR9921EA 24.87 USD
PAR9921
4-CORE® Buffer Pack, Promega®
Buffers Enzyme Buffers
Buffer pack contains convenient aliquots of Promega® restriction enzyme 10X buffers A, B, C, and D
The majority of Promega® restriction enzymes have optimal activity in one of these four 10X reaction buffers.
Información para pedidos: Pack contains 1 mL each of buffers A, B, C, and D.
Precaución: Note: For laboratory use. Outside of the United States, this product is intended for research use only, unless otherwise stated.