Bgl I (Genome Qualified), Restriction Enzymes, Promega

Supplier: Promega Corporation

R6071
PAR6071EA 96.92 USD
PAR6071
Bgl I (Genome Qualified), Restriction Enzymes, Promega
Nucleic Acid Reagents Restriction Enzymes

GCCN NNN/NGGC CGGN\NNN NCCG


  • Genome qualified to ensure optimal performance in genomic analysis applications

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme Name Bgl I (Genome Qualified)
Source
Buffer supplied with enzyme Buffer D
Buffer activity % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 25…% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1
Enzyme assay temperature 37°C
Storage Conditions –30...–10 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR