Jailbreak Column Heater for Thermo Easy-Spray Source
Supplier: PHOENIX S&T INC MS
PST-ESR-17U PST-ESR-36U
MSPP-PSTESR17UEA 4441.54 USD
MSPP-PSTESR17U MSPP-PSTESR36U
Jailbreak Column Heater for Thermo Easy-Spray Source
Heaters Column Heaters
Plug-and-Play directly into the Easy-Spray source and is supported by the trap-column bracket on the source.
  • Simple, flexible and low-cost
  • Temperature is controlled by the Easy-Spray source with the same precision as that of the OEM Easy-Spray columns.
The high voltage for nano spray comes from the source and is applied through a liquid junction - supply your own Tee or stainless steel union or purchase as a set.
Order Now
SPECIFICATIONS
Environmentally Preferable