Anti-AGMAT Rabbit Polyclonal Antibody
Supplier: CLOUD-CLONE CORP MS
PAJ664HU01
MSPP-PAJ664HU1EA 350 USD
MSPP-PAJ664HU1
Anti-AGMAT Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Agmatine Ureohydrolase (AGMAT), derived from recombinant AGMAT(Pro53~Ala151), is reactive with Human/Pig.
  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee
Agmatinase, which hydrolyzes agmatine to putrescine and urea, not only represents a potentially important mechanism for regulating the biological effects ofMore Product Information