Anti-MASP1 Rabbit Polyclonal Antibody
Supplier: CLOUD-CLONE CORP MS
PAB895MU01
MSPP-PAB895MU1EA 342 USD
MSPP-PAB895MU1
Anti-MASP1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Mannose Associated Serine Protease 1 (MASP1), derived from recombinant MASP1 (His25~Arg453), is reactive with Mouse/Human/Rat.
  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee
Mannose-associated serine protease 1 (MASP1) is involved in the lectin pathway of the complement system and is responsible for cleavingMore Product Information