Anti-MYL2 Rabbit Polyclonal Antibody
Supplier: CLOUD-CLONE CORP MS
PAB102HU01
MSPP-PAB102HU1EA 332 USD
MSPP-PAB102HU1
Anti-MYL2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Myosin Light Chain 2 (MYL2), derived from recombinant MYL2(Ala2~Ile159), is reactive with Human/Mouse/Rat/Pig.
  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee
Myosin regulatory light chain 2, ventricular/cardiac muscle isoform is the regulatory light chain associated with cardiac myosin beta (or slow) heavyMore Product Information