96 well Glass Bottom Plate, Cellvis
Supplier: CELLVIS, LLC MS
P96-1.5H-N
MSPP-P9615HNCS 383.08 USD
MSPP-P9615HN
96 well Glass Bottom Plate, Cellvis
Microplates Cell Culture Plates
Black polystyrene frame with high performance cover glass (0.170 ±0.005 mm), with lid, Individually packed.
  • Suitable for long term tissue culture
  • Manufactured in a class 100000 clean room
  • Plate made from virgin polystyrene, tissue culture treated
  • German high quality cover glass of superior optical quality, the cover glass has a thickness of 0.170 ±0.005 mm
  • Sterilized by gamma radiation
Suitable for Differential interference contrast (DIC), widefield fluorescence, confocal microscopy, two-photon and multiphoton microscopy, fluorescence recovery afterMore Product Information