Ultra-Low Temperature Chest Mini Freezer, MV85-2 Model, So-Low Environmental Equip MS
Supplier: SO-LOW ENVIRONMENTAL EQUIP MS
MV85-2
MSPP-MV85-2EA 12661.54 USD
MSPP-MV85-2
Ultra-Low Temperature Chest Mini Freezer, MV85-2 Model, So-Low Environmental Equip MS
Freezers Ultra Low Temperature Freezers
This ultra low temperature mini freezer ensures the safe storage of samples at –40 to −85 °C.
  • Digital control with data-logging capability
  • Battery operated back-up alarm, and Hi/Low alarm
  • Remote alarm hook-up is also provided
  • One year warranty
The freezer should be placed with at least 4" of space on sides and back. Battery operated alarm system will emit an audible and visual signal when there is a mechanical or electrical failure.
Certificaciones: Supplied by Nation-Wide-Network of service companies. ETL electrical approval.