Nitrogen Generator, Solaris 10 ELSD
Supplier: PEAK SCIENTIFIC MS
65-0001
MSPP-65-0001EA 10852.92 USD
MSPP-65-0001
Nitrogen Generator, Solaris 10 ELSD
Gas Generators Nitrogen Generators
Peak Scientific’s Solaris 10 Nitrogen generator has been developed to provide an optimal gas generation solution for ELSD or Compact Mass Spectrometers, with the goal of reducing instrument downtime and improving workflow efficiencies.
  • Variable outlet flow, pressure and purity
  • UHP nitrogen for ELSD at up to 10 L/min, 100 psi and up to 99.5% purity
  • Compact benchtop form factor, discrete aesthetic styling
  • Colour indicative LED lighting for easy status awareness
  • Optional Solaris Air Compressor stackable module
  • Engineered on reliable and proven existing
More Product Information