Hot Bead Sterilizers, Fine Science Tools
Supplier: FINE SCIENCE TOOLS INC MS
18000-45
MSPP-1800045EA 924.62 USD
MSPP-1800045
Hot Bead Sterilizers, Fine Science Tools
Sterilizers Glass Bead Sterilizers
The sterilizer provides a convenient method for sterilizing your surgical instruments prior to surgery or tissue culture. Without using potentially dangerous gases, flames or chemicals, you can ensure that your instruments will be free of pathogens and microbial contaminants.
  • Safe and effective
  • The sterilizer is safe to use even in laminar flow hoods
  • Well diameter: 40 mm
The beads take 15 to 20 minutes to reach 240 to 270 °C (464 to 518 °F), and then the sterilizer is ready for use. The temperature is thermostatically maintained, and the unit can beMore Product Information