Dionex™ IonPac™ AS19 IC Columns, Thermo Scientific
Supplier: THERMO ELECTRON NORTH AMERICA MS
062886 062888
MSPP-062886EA 2569.23 USD
MSPP-062886 MSPP-062888
Dionex™ IonPac™ AS19 IC Columns, Thermo Scientific
Chromatography Columns
This column is specifically designed and recommended for determination of trace bromate, a toxic byproduct of ozone disinfection.
  • Compatible with organic solvents
  • Low background signal and enhanced analyte sensitivity
  • Analyse most drinking water without the use of sample pretreatment or preconcentration
  • Operates at ambient or elevated temperatures with column selectivity optimised for 30 °C
  • Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300) and a Reagent-Free™ (RFIC™) system
The column's selectivity ensures that bromate can be quantified at low µg/More Product Information