Iodine 0.0282 N, StandARd® Volumetric (APHA), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIALS US


M.O.C. Category N

H169-05
MKH16905CS 422.19 USD
MKH16905
Iodine 0.0282 N, StandARd® Volumetric (APHA), Macron Fine Chemicals™
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 1.00 g/cm³
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8 (6.1) III
Merck Index: 13,05036

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0277 - 0.0287
Appearance (transparent, reddish-brown solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR