Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIALS US
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

Management of Change (MOC) category = N

7581-06 7581-12 7581-24 7581-60 7581-19 7581-20
MK758106CS 726.97 USD
MK758106 MK758112 MK758124 MK758160 MK758119 MK758120
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Sodium chloride

AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NaCl) (by Ag titrn) ≥99.0 %
ACS - pH of 5% Solution at 25°C 5.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Iodide (I) ≤0.002 %
ACS - Bromide (Br) ≤0.01 %
ACS - Chlorate and Nitrate (as NO₃) ≤0.003 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.004 %
ACS - Barium (Ba) Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤2 ppm
ACS - Calcium (Ca) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
Appearance (white, crystalline granules)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR