Masterflex® Single-Use Gamma Irradiated Sampling Bottle Assemblies, Sterile, Avantor®
Supplier: VWR International
MFLX95644-04 MFLX95644-00 MFLX95644-02 MFLX95644-06 MFLX95644-08
MFLX95644-04EA 107 USD
MFLX95644-04 MFLX95644-00 MFLX95644-02 MFLX95644-06 MFLX95644-08
Masterflex® Single-Use Gamma Irradiated Sampling Bottle Assemblies, Sterile, Avantor®
Bottles Plastic Bottles
Safely transfer, sample, and store biopharmaceutical products and reagents.
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain sterility until use. Vent tube includes a 0.2-micron PTFE ventMore Product Information