Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412 ISM4206 ISM4208 ISM4212 ISM4306 ISM4308 ISM4312 ISM4406 ISM4408 ISM4412
MFLX78001-58EA 4600 USD
MFLX78001-58 MFLX78001-60 MFLX78001-62 MFLX78001-68 MFLX78001-70 MFLX78001-72 MFLX78001-78 MFLX78001-80 MFLX78001-82 MFLX78001-58-AU MFLX78001-60-AU MFLX78001-62-AU MFLX78001-68-AU MFLX78001-70-AU MFLX78001-72-AU MFLX78001-78-AU MFLX78001-80-AU MFLX78001-82-AU MFLX78001-58-DK MFLX78001-60-DK MFLX78001-62-DK MFLX78001-68-DK MFLX78001-70-DK MFLX78001-72-DK MFLX78001-78-DK MFLX78001-80-DK MFLX78001-82-DK MFLX78001-58-IN MFLX78001-60-IN MFLX78001-62-IN MFLX78001-68-IN MFLX78001-70-IN MFLX78001-72-IN MFLX78001-78-IN MFLX78001-80-IN MFLX78001-82-IN MFLX78001-58-IL MFLX78001-60-IL MFLX78001-62-IL MFLX78001-68-IL MFLX78001-70-IL MFLX78001-72-IL MFLX78001-78-IL MFLX78001-80-IL MFLX78001-82-IL MFLX78001-58-EU MFLX78001-60-EU MFLX78001-62-EU MFLX78001-68-EU MFLX78001-70-EU MFLX78001-72-EU MFLX78001-78-EU MFLX78001-80-EU MFLX78001-82-EU MFLX78001-58-UK MFLX78001-60-UK MFLX78001-62-UK MFLX78001-68-UK MFLX78001-70-UK MFLX78001-72-UK MFLX78001-78-UK MFLX78001-80-UK MFLX78001-82-UK MFLX78001-58-CH MFLX78001-60-CH MFLX78001-62-CH MFLX78001-68-CH MFLX78001-70-CH MFLX78001-72-CH MFLX78001-78-CH MFLX78001-80-CH MFLX78001-82-CH MFLX78001-58-IT MFLX78001-60-IT MFLX78001-62-IT MFLX78001-68-IT MFLX78001-70-IT MFLX78001-72-IT MFLX78001-78-IT MFLX78001-80-IT MFLX78001-82-IT
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensingMore Product Information