Masterflex® I/P® Digital Precision Modular Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50 77970-40 77970-50
MFLX77970-40EA 7420 USD
MFLX77970-40 MFLX77970-50 MFLX77970-40-AU MFLX77970-50-AU MFLX77970-40-DK MFLX77970-50-DK MFLX77970-40-IN MFLX77970-50-IN MFLX77970-40-IL MFLX77970-50-IL MFLX77970-40-EU MFLX77970-50-EU MFLX77970-40-UK MFLX77970-50-UK MFLX77970-40-CH MFLX77970-50-CH MFLX77970-40-IT MFLX77970-50-IT
Masterflex® I/P® Digital Precision Modular Drive Systems, Avantor®
Pumps Peristaltic Pumps
Place motor and controller where convenient with modular design.
  • Flow rate 0.0006 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L,
More Product Information