Masterflex® I/P® MasterSense® Process Pumps with I/P® Rapid-Load® Pump Head
Supplier: Avantor Fluid Handling
77966-20 77966-22 77966-30 77966-32 77966-20-AU 77966-22-AU 77966-30-AU 77966-32-AU 77966-20-DK 77966-22-DK 77966-30-DK 77966-32-DK 77966-20-IN 77966-22-IN 77966-30-IN 77966-32-IN 77966-20-EU 77966-22-EU 77966-30-EU 77966-32-EU 77966-20-UK 77966-22-UK 77966-30-UK 77966-32-UK 77966-20-CH 77966-22-CH 77966-30-CH 77966-32-CH 77966-20-IT 77966-22-IT 77966-30-IT 77966-32-IT 77966-20-IL 77966-22-IL 77966-30-IL 77966-32-IL
MFLX77966-20EA 14473 USD
MFLX77966-20 MFLX77966-22 MFLX77966-30 MFLX77966-32 MFLX77966-20-AU MFLX77966-22-AU MFLX77966-30-AU MFLX77966-32-AU MFLX77966-20-DK MFLX77966-22-DK MFLX77966-30-DK MFLX77966-32-DK MFLX77966-20-IN MFLX77966-22-IN MFLX77966-30-IN MFLX77966-32-IN MFLX77966-20-EU MFLX77966-22-EU MFLX77966-30-EU MFLX77966-32-EU MFLX77966-20-UK MFLX77966-22-UK MFLX77966-30-UK MFLX77966-32-UK MFLX77966-20-CH MFLX77966-22-CH MFLX77966-30-CH MFLX77966-32-CH MFLX77966-20-IT MFLX77966-22-IT MFLX77966-30-IT MFLX77966-32-IT MFLX77966-20-IL MFLX77966-22-IL MFLX77966-30-IL MFLX77966-32-IL
Masterflex® I/P® MasterSense® Process Pumps with I/P® Rapid-Load® Pump Head
Pumps Peristaltic Pumps
Versatile pumps for process applications - with an intuitive touch-screen interface and advanced connectivity. This drive offers a broad flow range, from 0.0006 to 19 LPM, adaptable to a variety of applications and workflows. The drive housing is constructed of 316 stainless steel - IP66 and NEMA 4X rated for washdown.
  • Powerful, space-efficient drives will accept all I/P® pump head and tubing options
  • I/P® Rapid-Load® pump head offers the best combination of pressure and flow performance
  • 17.8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and
More Product Information