Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
77919-25 77919-35 77921-65 77921-75 77921-77 77921-85 77919-35 77921-65 77921-75 77921-85 77919-25 77919-35 77921-65 77921-75 77921-85 77919-25 77919-35 77921-65 77921-75 77921-85 77919-25 77919-35 77921-65 77921-75 77921-85 77919-25 77921-65 77921-75 77921-85 77919-25 77921-65 77921-75 77921-85 77919-25 77919-35 77921-65 77921-75 77921-85 77919-25 77919-35 77921-65 77921-75 77921-85
MFLX77919-25EA 5910 USD
MFLX77919-25 MFLX77919-35 MFLX77921-65 MFLX77921-75 MFLX77921-77 MFLX77921-79 MFLX77921-85 MFLX77919-25-AU MFLX77919-35-AU MFLX77921-65-AU MFLX77921-75-AU MFLX77921-77-AU MFLX77921-79-AU MFLX77921-85-AU MFLX77919-25-DK MFLX77919-35-DK MFLX77921-65-DK MFLX77921-75-DK MFLX77921-77-DK MFLX77921-79-DK MFLX77921-85-DK MFLX77919-25-IN MFLX77919-35-IN MFLX77921-65-IN MFLX77921-75-IN MFLX77921-77-IN MFLX77921-79-IN MFLX77921-85-IN MFLX77919-25-IL MFLX77919-35-IL MFLX77921-65-IL MFLX77921-75-IL MFLX77921-77-IL MFLX77921-79-IL MFLX77921-85-IL MFLX77919-25-EU MFLX77919-35-EU MFLX77921-65-EU MFLX77921-75-EU MFLX77921-77-EU MFLX77921-79-EU MFLX77921-85-EU MFLX77919-25-UK MFLX77919-35-UK MFLX77921-65-UK MFLX77921-75-UK MFLX77921-77-UK MFLX77921-79-UK MFLX77921-85-UK MFLX77919-25-CH MFLX77919-35-CH MFLX77921-65-CH MFLX77921-75-CH MFLX77921-77-CH MFLX77921-79-CH MFLX77921-85-CH MFLX77919-25-IT MFLX77919-35-IT MFLX77921-65-IT MFLX77921-75-IT MFLX77921-77-IT MFLX77921-79-IT MFLX77921-85-IT
Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Pumps Peristaltic Pumps
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures
More Product Information