Masterflex® I/P® Brushless Process Drives, Avantor®
Supplier: Avantor Fluid Handling
77410-10 77714-18 77714-31 77410-10 77410-10 77410-10 77410-10 77410-10 77410-10 77410-10 77410-10
MFLX77410-10EA 4310 USD
MFLX77410-10 MFLX77714-18 MFLX77714-31 MFLX77410-10-AU MFLX77410-10-DK MFLX77410-10-IN MFLX77410-10-IL MFLX77410-10-EU MFLX77410-10-UK MFLX77410-10-CH MFLX77410-10-IT
Masterflex® I/P® Brushless Process Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Powerful, low-maintenance drives are versatile and cost-effective.
  • Flow rate 0.2 to 19 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
I/P® brushless process drives offer heavy-More Product Information