Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®
Supplier: Avantor Fluid Handling
77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70 77301-40 77301-50 77301-60 77301-70
MFLX77301-40EA 4640 USD
MFLX77301-40 MFLX77301-50 MFLX77301-60 MFLX77301-70 MFLX77301-40-AU MFLX77301-50-AU MFLX77301-60-AU MFLX77301-70-AU MFLX77301-40-DK MFLX77301-50-DK MFLX77301-60-DK MFLX77301-70-DK MFLX77301-40-IN MFLX77301-50-IN MFLX77301-60-IN MFLX77301-70-IN MFLX77301-40-IL MFLX77301-50-IL MFLX77301-60-IL MFLX77301-70-IL MFLX77301-40-EU MFLX77301-50-EU MFLX77301-60-EU MFLX77301-70-EU MFLX77301-40-UK MFLX77301-50-UK MFLX77301-60-UK MFLX77301-70-UK MFLX77301-40-CH MFLX77301-50-CH MFLX77301-60-CH MFLX77301-70-CH MFLX77301-40-IT MFLX77301-50-IT MFLX77301-60-IT MFLX77301-70-IT
Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.
  • Flow rate 0.0012 to 3400 ml/min with L/S —flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for
More Product Information