Masterflex® L/S® Cytoflow™ Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
77253-00 77253-02 77253-10 77253-12
MFLX77253-00EA 1420 USD
MFLX77253-00 MFLX77253-02 MFLX77253-10 MFLX77253-12
Masterflex® L/S® Cytoflow™ Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
The best option for pumping live cells and shear-sensitive fluids—validated by independent test data.
  • Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
  • Large-diameter rotor delivers higher flow rates at lower speeds
  • Ensure the survival and vitality of live cells
  • Flow rate 0.03 to 5700 ml/min with L/S tubing—flow rate depends on the drive rpm, pump head, and tubing size
  • Compatible with all Masterflex L/S drives that accept two or more pump heads
  • Models available for Masterflex L/S Precision and L/S
More Product Information