Masterflex® Ismatec® Microflex Variable-Speed Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34 77122-30 77122-32 77122-34
MFLX77122-30EA 1280 USD
MFLX77122-30 MFLX77122-32 MFLX77122-34 MFLX77122-30-AU MFLX77122-32-AU MFLX77122-34-AU MFLX77122-30-DK MFLX77122-32-DK MFLX77122-34-DK MFLX77122-30-IN MFLX77122-32-IN MFLX77122-34-IN MFLX77122-30-IL MFLX77122-32-IL MFLX77122-34-IL MFLX77122-30-EU MFLX77122-32-EU MFLX77122-34-EU MFLX77122-30-UK MFLX77122-32-UK MFLX77122-34-UK MFLX77122-30-CH MFLX77122-32-CH MFLX77122-34-CH MFLX77122-30-IT MFLX77122-32-IT MFLX77122-34-IT
Masterflex® Ismatec® Microflex Variable-Speed Pump Systems, Avantor®
Pumps Peristaltic Pumps
Easy-to-use low-flow pumps feature reversible pumping, remote start/stop, and rapid tubing changes.
  • Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
  • Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings.
  • Separate single-turn speed control and cw/off/ccw switch with green LED power indicator—maintain speed setting when you turn pump on/off or reverse direction.
  • Reversible permanent magnet DC motor; purge fluid before or after pumping; pump fluid in either
More Product Information