Masterflex® Single-Use Replacement Pump Heads with EZ-Set for Quattroflow™ Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
76000-01 76000-03
MFLX76000-01EA 1220 USD
MFLX76000-01 MFLX76000-03
Masterflex® Single-Use Replacement Pump Heads with EZ-Set for Quattroflow™ Pump Systems, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
  • EZ-Set design allows for quick pump head change—reducing downtime
  • Constructed with a disposable polypropylene chamber—ideal for single-use applications
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
The Masterflex® Quattroflow™ single-use replacement pump heads offer a unique four diaphragm (chamber) design,More Product Information