Micropump® B-Mount Gear Pump Heads
Supplier: MICROPUMP INC
MI0013 / Z-120 / 82004 MI0019 / Z-130 / 81529 MI0267 / Z-040 / 81977 MI0016 / Z-140 / 82001 MI0018 / Z-142 / 82003 MI0020 / Z-150 / 81531 MI0006 / Z-186 / 82092 MI0278 / Z-186 PPS / L18412 MI0007 / Z-181 / 82114 MI0008 / Z-183 / 82115 MI0131 / Z-1830 / 82004 MI0312 / Z-186P / L20820 MI0280 / Z-1830P / L18489 MI0313 / Z-140P / L19735 82006
MFLX74004-01CS 926.75 USD
MFLX74004-01 MFLX74004-05 MFLX74004-07 MFLX74004-09 MFLX74004-11 MFLX74004-17 MFLX74004-23 MFLX74004-27 MFLX74004-29 MFLX74004-31 MFLX74004-33 MFLX74004-71 MFLX74004-73 MFLX74004-75 MFLX74004-03 MFLX74004-55 MFLX74004-13 MFLX74004-53
Micropump® B-Mount Gear Pump Heads
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads areMore Product Information