Masterflex® Adapter Fittings, Hosebarb to NPT(M), Straight, Polyproylene, Avantor®
Supplier: VWR International
MFLX06365-42 MFLX30615-71 MFLX31407-16 MFLX40610-36 MFLX40610-38 MFLX40610-39 MFLX40610-40 MFLX40610-41 MFLX40610-42 MFLX40610-43 MFLX40610-44 MFLX40610-45 MFLX40610-46 MFLX40610-47 MFLX40610-48 MFLX40610-50 MFLX40610-53 MFLX40621-11 MFLX40621-22 MFLX40621-25 MFLX40621-28 MFLX40621-41 MFLX40621-45 MFLX40621-51 MFLX40621-61 MFLX40621-69 MFLX40625-28 MFLX40625-45 MFLX40625-61 MFLX40640-36 MFLX40640-37 MFLX40640-39 MFLX40640-45 MFLX41517-27 MFLX41517-47 MFLX30615-51 MFLX30615-63 MFLX30615-67 MFLX31407-12 MFLX31407-13 MFLX31407-14 MFLX31407-19 MFLX31407-20 MFLX40610-49 MFLX40610-51 MFLX40610-52 MFLX40625-41 MFLX40640-38 MFLX40640-41 MFLX40640-42 MFLX40640-44 MFLX40640-46 MFLX40640-51 MFLX40625-51 MFLX40640-43 MFLX40640-50 MFLX40610-37
MFLX06365-42CS 0 USD
MFLX06365-42 MFLX30615-71 MFLX31407-16 MFLX40610-36 MFLX40610-38 MFLX40610-39 MFLX40610-40 MFLX40610-41 MFLX40610-42 MFLX40610-43 MFLX40610-44 MFLX40610-45 MFLX40610-46 MFLX40610-47 MFLX40610-48 MFLX40610-50 MFLX40610-53 MFLX40621-11 MFLX40621-22 MFLX40621-25 MFLX40621-28 MFLX40621-41 MFLX40621-45 MFLX40621-51 MFLX40621-61 MFLX40621-69 MFLX40625-28 MFLX40625-45 MFLX40625-61 MFLX40640-36 MFLX40640-37 MFLX40640-39 MFLX40640-45 MFLX41517-27 MFLX41517-47 MFLX30615-51 MFLX30615-63 MFLX30615-67 MFLX31407-12 MFLX31407-13 MFLX31407-14 MFLX31407-19 MFLX31407-20 MFLX40610-49 MFLX40610-51 MFLX40610-52 MFLX40625-41 MFLX40640-38 MFLX40640-41 MFLX40640-42 MFLX40640-44 MFLX40640-46 MFLX40640-51 MFLX40625-51 MFLX40640-43 MFLX40640-50 MFLX40610-37
Masterflex® Adapter Fittings, Hosebarb to NPT(M), Straight, Polyproylene, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in aMore Product Information