Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
Supplier: VWR International
MFLX30703-08 MFLX40614-39 MFLX40614-40 MFLX40614-41 MFLX40614-42 MFLX40614-43 MFLX40614-44 MFLX40614-45 MFLX40614-46 MFLX40614-47 MFLX40614-48 MFLX40614-49 MFLX40614-50 MFLX40614-38
MFLX30703-08EA 62.3 USD
MFLX30703-08 MFLX40614-39 MFLX40614-40 MFLX40614-41 MFLX40614-42 MFLX40614-43 MFLX40614-44 MFLX40614-45 MFLX40614-46 MFLX40614-47 MFLX40614-48 MFLX40614-49 MFLX40614-50 MFLX40614-38
Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
Fittings and Connectors
Quick and easy assembly without tools.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Ultra smooth inner surface results in fewer extractables
These economicalMore Product Information