Masterflex® Paddle Wheel Flowmeters for Monitoring and Control, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PWE04PELNB2 PWE06PELRB2 PWE010PELRB2 MFLX32755-17 MFLX32755-18 MFLX32755-24 MFLX32755-32 MFLX32755-34 MFLX32755-35 MFLX32755-44 MFLX32755-45 MFLX32755-46
MFLX32755-31EA 874 USD
MFLX32755-31 MFLX32755-36 MFLX32755-38 MFLX32755-17 MFLX32755-18 MFLX32755-24 MFLX32755-32 MFLX32755-34 MFLX32755-35 MFLX32755-44 MFLX32755-45 MFLX32755-46
Masterflex® Paddle Wheel Flowmeters for Monitoring and Control, Avantor®
Flow Meters
Meters provide visual and remote flow indication for numerous applications.
  • Choose from standard or digital models with display and keypad
  • Manufactured from solid PP or PVDF; choose PVDF for better chemical resistance
  • Jewel bearings allow for very low minimum flow rates
  • Only one moving part provides extended life
Designed for water-based liquids, these IP65-rated flowmeters provide flow measurement to 35 GPM (132.5 LPM) and models with the RTD option measure temperature to 140 °F (60 °C). The RTD is platinum encased in 316 stainless steel and has a 4-More Product Information