Masterflex® I/P® Digital Washdown Modular Drive with Remote Input/Output, Avantor®
Supplier: Avantor Fluid Handling
7591-30 7591-30 7591-30 7591-30 7591-30 7591-30 7591-30 7591-30 7591-30
MFLX07591-30EA 5510 USD
MFLX07591-30 MFLX07591-30-AU MFLX07591-30-DK MFLX07591-30-IN MFLX07591-30-IL MFLX07591-30-EU MFLX07591-30-UK MFLX07591-30-CH MFLX07591-30-IT
Masterflex® I/P® Digital Washdown Modular Drive with Remote Input/Output, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Place motor and controller where convenient with modular design.
  • Flow rate 0.04 to 19 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows pump rpm— confirm speed setting at a glance
  • Remote control available using analog communication—voltage, current and contact closure
  • Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
  • Membrane keypad allows for easy up-front control of speed, direction, and start/stop
  • Rugged housing is NEMA 4
More Product Information