Masterflex® L/S® Economy Fixed-Speed Drives, Avantor®
Supplier: Avantor Fluid Handling
7540-01 7540-02 7540-06 7540-12 7540-20 7540-30 7540-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60 7542-01 7542-02 7542-06 7542-12 7542-20 7542-30 7542-60
MFLX07540-01EA 579 USD
MFLX07540-01 MFLX07540-02 MFLX07540-06 MFLX07540-12 MFLX07540-20 MFLX07540-30 MFLX07540-60 MFLX07542-01 MFLX07542-02 MFLX07542-06 MFLX07542-12 MFLX07542-20 MFLX07542-30 MFLX07542-60 MFLX07542-01-AU MFLX07542-02-AU MFLX07542-06-AU MFLX07542-12-AU MFLX07542-20-AU MFLX07542-30-AU MFLX07542-60-AU MFLX07542-01-DK MFLX07542-02-DK MFLX07542-06-DK MFLX07542-12-DK MFLX07542-20-DK MFLX07542-30-DK MFLX07542-60-DK MFLX07542-01-IN MFLX07542-02-IN MFLX07542-06-IN MFLX07542-12-IN MFLX07542-20-IN MFLX07542-30-IN MFLX07542-60-IN MFLX07542-01-IL MFLX07542-02-IL MFLX07542-06-IL MFLX07542-12-IL MFLX07542-20-IL MFLX07542-30-IL MFLX07542-60-IL MFLX07542-01-EU MFLX07542-02-EU MFLX07542-06-EU MFLX07542-12-EU MFLX07542-20-EU MFLX07542-30-EU MFLX07542-60-EU MFLX07542-01-UK MFLX07542-02-UK MFLX07542-06-UK MFLX07542-12-UK MFLX07542-20-UK MFLX07542-30-UK MFLX07542-60-UK MFLX07542-01-CH MFLX07542-02-CH MFLX07542-06-CH MFLX07542-12-CH MFLX07542-20-CH MFLX07542-30-CH MFLX07542-60-CH MFLX07542-01-IT MFLX07542-02-IT MFLX07542-06-IT MFLX07542-12-IT MFLX07542-20-IT MFLX07542-30-IT MFLX07542-60-IT
Masterflex® L/S® Economy Fixed-Speed Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Economical drives for reliable general transfer and feed applications.
  • Economy fixed-speed drives are stackable and ideal for lab, field, or process feed applications
  • Choose from seven speeds based on flow rate or number of heads required
  • Constructed with painted steel housing for IP22-rating
The Masterflex® L/S® economy fixed-speed drives are compact and ideal where space is limited. These drives feature a lighted power switch on the front of the drive that lets you know when the pump is running. They accept five different pump head types: Standard,More Product Information