Masterflex® L/S® Precision Process Drives, Avantor®
Supplier: Avantor Fluid Handling
7528-40 7528-50 7528-40 MFLX07528-45-AU 7528-50 7528-40 MFLX07528-45-DK 7528-50 7528-40 MFLX07528-45-IN 7528-50 7528-40 MFLX07528-45-IL 7528-50 7528-40 MFLX07528-45-EU 7528-50 7528-40 MFLX07528-45-UK 7528-50 7528-40 MFLX07528-45-CH 7528-50 7528-40 MFLX07528-45-IT 7528-50 MFLX07528-55-AU MFLX07528-55-DK MFLX07528-55-IN MFLX07528-55-IL MFLX07528-55-EU MFLX07528-55-UK MFLX07528-55-CH MFLX07528-55-IT
MFLX07528-40EA 4480 USD
MFLX07528-40 MFLX07528-45 MFLX07528-50 MFLX07528-40-AU MFLX07528-45-AU MFLX07528-50-AU MFLX07528-40-DK MFLX07528-45-DK MFLX07528-50-DK MFLX07528-40-IN MFLX07528-45-IN MFLX07528-50-IN MFLX07528-40-IL MFLX07528-45-IL MFLX07528-50-IL MFLX07528-40-EU MFLX07528-45-EU MFLX07528-50-EU MFLX07528-40-UK MFLX07528-45-UK MFLX07528-50-UK MFLX07528-40-CH MFLX07528-45-CH MFLX07528-50-CH MFLX07528-40-IT MFLX07528-45-IT MFLX07528-50-IT MFLX07528-55 MFLX07528-55-AU MFLX07528-55-DK MFLX07528-55-IN MFLX07528-55-IL MFLX07528-55-EU MFLX07528-55-UK MFLX07528-55-CH MFLX07528-55-IT
Masterflex® L/S® Precision Process Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Flow rate 0.06 to 3400 ml/min (depends on drive rpm and tubing size)
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for
More Product Information