Masterflex® L/S® Standard Digital Drives, Avantor®
Supplier: Avantor Fluid Handling
7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30 7522-20 7522-30
MFLX07522-20EA 3670 USD
MFLX07522-20 MFLX07522-30 MFLX07522-38 MFLX07522-20-AU MFLX07522-28-AU MFLX07522-30-AU MFLX07522-38-AU MFLX07522-20-DK MFLX07522-28-DK MFLX07522-30-DK MFLX07522-38-DK MFLX07522-20-IN MFLX07522-28-IN MFLX07522-30-IN MFLX07522-38-IN MFLX07522-20-IL MFLX07522-28-IL MFLX07522-30-IL MFLX07522-38-IL MFLX07522-20-EU MFLX07522-28-EU MFLX07522-30-EU MFLX07522-38-EU MFLX07522-20-UK MFLX07522-28-UK MFLX07522-30-UK MFLX07522-38-UK MFLX07522-20-CH MFLX07522-28-CH MFLX07522-30-CH MFLX07522-38-CH MFLX07522-20-IT MFLX07522-28-IT MFLX07522-30-IT MFLX07522-38-IT
Masterflex® L/S® Standard Digital Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Flow rate 0.001/0.006 to 566/3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles;
More Product Information