FMI Valveless Piston Pump Heads, Series Q
Supplier: FLUID METERING INC
FMI217 / Q3CKC FMI352 / Q1CKC + (2) R412-2K FMI205 / Q1CSC FMI462 / Q3CSC FMI355 / Q2CKC Q2CSC
MFLX07104-50EA 1160 USD
MFLX07104-50 MFLX07104-52 MFLX07104-53 MFLX07104-54 MFLX07104-56 MFLX07104-58 MFLX07104-62 MFLX07104-63 MFLX07104-64 MFLX07104-66 MFLX07104-70 MFLX07104-59 MFLX07104-60 MFLX07104-68 MFLX07104-72 MFLX07117-26 MFLX07117-28
FMI Valveless Piston Pump Heads, Series Q
Pumps Pump Heads Piston Pump Heads
No valves to clog or wear out.
  • Build your own pump systems using interchangeable heads, drives, and accessories
  • Pump fluids with viscosities up to 500 cp
  • Low dead volume
These FMI Series Q valveless piston pump heads give you options to handle a wide range of fluids at pressures up to 100 psi. The interchangeable pump head modules provide numerous flow and material combinations for optimal versatility. Choose from a combination of ceramic, PVDF, carbon, and stainless steel wetted materials for an inert and corrosion resistant fluid path, ideal forMore Product Information