Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-G
Supplier: VWR International
MFLX06404-13 MFLX06404-14 MFLX06404-16 MFLX06404-17 MFLX06404-18 MFLX06404-25
MFLX06404-13CS 83.9 USD
MFLX06404-13 MFLX06404-14 MFLX06404-16 MFLX06404-17 MFLX06404-18 MFLX06404-25
Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-G
Tubing Peristaltic Pump Tubing
Well-suited for vacuum/pressure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-
More Product Information