Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
T51/1-N032-41G T51/1-N032-41S T51/1-N062-01S T51/1-N112-02S T51/1-N082-03S T51/1-N092-04S P01/1-N032-41ST P01/1-N062-01ST P01/1-N082-03G P01/1-N082-03ST P01/1-N092-04ST P01/1-N102-05ST P01/1-N034-39G P01/1-N034-39ST P21/1-N032-41ST P21/1-N062-01ST P21/1-N112-02ST P21/1-N082-03G P21/1-N082-03ST P21/1-N092-04G P21/1-N092-04ST P21/1-N102-05ST P21/1-N034-39G P21/1-N034-39ST P11/1-N062-01ST P11/1-N082-03G P11/1-N082-03ST T51/1-N062-01G T51/1-N112-02G T51/1-082-03G T51/1-N102-05S T51/1-N034-39S T51/1-092-04G P01/1-N032-41G P01/1-N062-01G P01/1-N112-02G P01/1-N112-02ST P01/1-N092-04G P01/1-N102-05G T51/1-034-39G P21/1-N032-41G P21/1-N062-01G P21/1-N112-02G P21/1-N102-05G P11/1-N032-41G P11/1-N032-41ST P11/1-N062-01G P11/1-N112-02G P11/1-N112-02ST P11/1-N092-04G P11/1-N092-04ST P11/1-N102-05G P11/1-N102-05ST P11/1-N034-39G
MFLX03217-52EA 471 USD
MFLX03217-52 MFLX03217-53 MFLX03217-57 MFLX03217-62 MFLX03217-67 MFLX03217-72 MFLX03217-73 MFLX03227-00 MFLX03227-06 MFLX03227-10 MFLX03227-16 MFLX03227-18 MFLX03227-22 MFLX03227-26 MFLX03227-28 MFLX03227-30 MFLX03229-01 MFLX03229-03 MFLX03229-07 MFLX03229-11 MFLX03229-15 MFLX03229-17 MFLX03229-19 MFLX03229-21 MFLX03229-23 MFLX03229-27 MFLX03229-29 MFLX03229-31 MFLX03295-10 MFLX03295-16 MFLX03295-18 MFLX03217-58 MFLX03217-63 MFLX03217-68 MFLX03217-74 MFLX03217-76 MFLX03217-79 MFLX03227-02 MFLX03227-04 MFLX03227-08 MFLX03227-12 MFLX03227-14 MFLX03227-20 MFLX03227-24 MFLX03227-84 MFLX03229-05 MFLX03229-09 MFLX03229-13 MFLX03229-25 MFLX03295-00 MFLX03295-02 MFLX03295-04 MFLX03295-06 MFLX03295-08 MFLX03295-12 MFLX03295-14 MFLX03295-20 MFLX03295-22 MFLX03295-24 MFLX03295-26 MFLX03295-28
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information