Z-Lys-ONp hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-4-Nitrophenyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride, Z-Lys-ONp·HCl, Z-Lys-ONp HCl

4001257.0250 4001257.1000 4001257.5000
M-1295.0250BAEA 114.5 USD
M-1295.0250BA M-1295.1000BA M-1295.5000BA
Z-Lys-ONp hydrochloride
Z-Lys-ONp hydrochloride

Sequence: Z-Lys-ONp · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


A synthetic chromogenic substrate for the determination of serinic and thiol proteases, e.g. urokinase, trypsin, cathepsin B, cathepsin L, and papain.

Formula: C₂₀H₂₄ClN₃O₆
MW: 437.877 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039055
CAS Number: 2179-15-9
EINECS: 218-546-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR