ULTRA-WARE® Filtration Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
953915-0047
KT953915-0047EA 843.62 USD
KT953915-0047
ULTRA-WARE® Filtration Caps, Kimble Chase
Closures Safety Caps and Safety Waste Caps
These Filtration caps made of glass-filled PTFE with a vacuum adapter for 1/4” ID tubing.
  • Upper screw clamp holds a solvent pickup adapter or funnel
  • Coarse fritted glass filter support is removable
Información de suministro: Supplied with a PTFE or propylene o-ring.