KONTES® Four-Neck Reaction Vessel Lids, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
613000-0021
KT613000-4524CS 660.77 USD
KT613000-4524 KT613000-0021
KONTES® Four-Neck Reaction Vessel Lids, Kimble Chase
Lids Reaction Vessel Lids
For use with cylindrical or spherical reaction vessels with a 10.2cm (4") I.D
flange. Available with two standard side necks and one thermometer neck or three standard side necks. Thermometer neck [ST] joint: 10/30.
Información para pedidos: For clamps, additional flange sizes, and additional [ST] joint numbers, contact your Avantor sales representative.
Order Now
SPECIFICATIONS
Environmentally Preferable