KIMBLE® Barrett Distilling Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
610900-0200 610910-0125
KT610900-0200CS 1939.48 USD
KT610900-0200 KT610910-0125
KIMBLE® Barrett Distilling Flasks, DWK Life Sciences
Flasks Distillation Flasks
These flasks feature smooth, consistent, heavy walls for uniform heating and minimal breakage.
  • Designed to ASTM E133 specifications
  • Manufactured from 33 expansion, low extractable borosilicate glass
610910-0125 conforms to Flask B in ASTM E133.
610900-0200 conforms to Flask C in ASTM E133 and also is compatible for use with ASTM D1078.
Certificaciones: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Información para pedidos: Flask is supplied with one cork for the sidearm.
Order Now
SPECIFICATIONS
Environmentally Preferable