KIMBLE® Rotary Evaporator Flasks with Standard Joint Bead, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
608675-0224 608675-0324 608675-0624
KT608675-0124CS 60.37 USD
KT608675-0124 KT608675-0224 KT608675-0324 KT608675-0624
KIMBLE® Rotary Evaporator Flasks with Standard Joint Bead, DWK Life Sciences
Flasks Rotary Evaporator Flasks
Single neck flask with a standard taper outer joint designed for easy recovery of reaction products.
  • Heavy wall
  • Total contents are accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Manufactured from 33 expansion, low extractable borosilicate glass
Even wall thickness and minimum run-out make these flasks ideal for use with rotary evaporators.
Certificaciones: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.