KIMBLE® TLC Plate Streakers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
416430-0000
KT416430-0000EA 704.54 USD
KT416430-0000
KIMBLE® TLC Plate Streakers, DWK Life Sciences
TLC Plate Applicators
A simple device for uniformly streaking TLC plates.
  • Unit consists of a stainless steel base with an adjustable holder for a glass reservoir
  • Reference: Journal of Chemical Education, volume 47, No. 5, May 1970, page 404, "A simple streaker for preparative layer chromatography," L.J. Altman, James R.Trudell
  • Glassware manufactured from 33 expansion, low extractable borosilicate glass
To operate, the streaker is placed over the edge of the counter top and the plate to be streaked is positioned parallel to it. The bent capillary tip is then adjusted soMore Product Information