Trifluoroacetic acid 99,0, BAKER ANALYZED®

Danger

Synonyms: TFA

M.O.C. Category N

W729-05
JTW729-7CS 7699.86 USD
JTW729-7 JTW729-5
Trifluoroacetic acid 99,0, BAKER ANALYZED®
Trifluoroacetic acid

Lot analysis on label.

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (F₃CCOOH) ≥99.0 %
Residue after Evaporation ≤0.0100 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR