Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

Synonyms: Pimelic Ketone

Management of Change (MOC) category = N

9210-01 9210-03
JT9210-1CS 2483.04 USD
JT9210-1 JT9210-3
Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Cyclohexanone

Lot analysis on label.

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3 III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₀O) (by GC) ≥99.0 %
Color (APHA) ≤10
Density (g/mL) at 25°C 0.940 - 0.943
Residue after Evaporation ≤0.0500 %
Acidity (meq/g) ≤0.003
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Identification (by IR) Passes Test
NOTE: Color reported is at time of packaging; material will darken with age.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR