Sulfuric acid 93-98%, ULTREX® II, Ultrapure, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger


Management of Change (MOC) category = N

6902-05
JT6902-5EA 797.99 USD
JT6902-5
Sulfuric acid 93-98%, ULTREX® II, Ultrapure, J.T.Baker®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₂SO₄)(w/w) 93 - 98 %
Trace Impurities - Aluminum (Al) ≤50 ppt
Trace Impurities - Antimony (Sb) ≤50 ppt
Trace Impurities - Arsenic (As) ≤500 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤50 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Dysprosium (Dy) ≤10 ppt
Trace Impurities - Erbium (Er) ≤10 ppt
Trace Impurities - Europium (Eu) ≤10 ppt
Trace Impurities - Gadolinium (Gd) ≤10 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤100 ppt
Trace Impurities - Hafnium (Hf) ≤10 ppt
Trace Impurities - Holmium (Ho) ≤10 ppt
Trace Impurities - Indium (In) ≤10 ppt
Trace Impurities - Iron (Fe) ≤50 ppt
Trace Impurities - Lanthanum (La) ≤10 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤10 ppt
Trace Impurities - Magnesium (Mg) ≤50 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Mercury (Hg) ≤100 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤10 ppt
Trace Impurities - Nickel (Ni) ≤50 ppt
Trace Impurities - Niobium (Nb) ≤10 ppt
Trace Impurities - Palladium (Pd), For Information Only
Trace Impurities - Platinum (Pt), For Information Only
Trace Impurities - Potassium (K) ≤50 ppt
Trace Impurities - Praseodymium (Pr) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤50 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Samarium (Sm) ≤10 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se) ≤500 ppt
Trace Impurities - Silver (Ag) ≤50 ppt
Trace Impurities - Sodium (Na) ≤50 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Tantalum (Ta), For Information Only
Trace Impurities - Tellurium (Te) ≤50 ppt
Trace Impurities - Terbium (Tb) ≤10 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤10 ppt
Trace Impurities - Thulium (Tm) ≤10 ppt
Trace Impurities - Tin (Sn) ≤50 ppt
Trace Impurities - Titanium (Ti) ≤50 ppt
Trace Impurities - Tungsten (W) ≤10 ppt
Trace Impurities - Uranium (U) ≤10 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤10 ppt
Trace Impurities - Yttrium (Y) ≤10 ppt
Trace Impurities - Zinc (Zn) ≤50 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR