Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

Synonyms: Caustic soda

Management of Change (MOC) category = N

3727-R 3727-03 3727-01 3727-07 3727-09 3727-25
JT3727-REA 2420.36 USD
JT3727-R JT3727-3 JT3727-1 JT3727-7 JT3727-9 JT3727-25
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Sodium hydroxide

Material will freeze if stored below 16°C (60°F). Protect from air to avoid absorption of carbon dioxide. Lot analysis on label.


Precaución: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Assay (NaOH) (by acidimetry) 50 - 52 %
Color (APHA) ≤20
Sodium Carbonate (Na₂CO₃) ≤0.1 %
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.001 %
Ammonium Hydroxide Precipitate ≤0.01 %
Silica (SiO₂), For Information Only
Heavy Metals (as Ag) ≤0.001 %
Potassium (K) ≤0.01 %
Nitrogen Compounds (as N) ≤5 ppm
Phosphate (PO₄) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Mercury (Hg), For Information Only
Trace Impurities - Nickel (Ni) ≤5.000 ppm
IMPORTANT: Material will freeze if stored below 60°F (16°C).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR